Oakton® pH 150 Waterproof Portable pH/mV/Temperature Meter
Rugged, accurate, reliable – Best value for an IP67-rated waterproof meter!
  • Intuitive, user-friendly icons—electrode status displays pH slope
  • Grip-Clip™ holder quickly and easily secures the electrode to your beaker or container
  • Expanded data logging for up to 150 data sets
  • mV offset for calibration of ORP standards, zeroing mV value, and pH diagnostics
  • 700 hours battery-life or optional universal power adapter
  • Built-in stand and wall mount option allow for easy benchtop measurement or storage
£506.74 - £780.86GBP / Each
£608.09 - £937.03GBP (incl VAT)
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Waterproof Portable Meter with SJ All-in-One Electrode
£636.44GBP / Each
£763.73GBP (incl VAT)
COMPARE
Waterproof Portable Meter; Meter Only
£506.74GBP / Each
£608.09GBP (incl VAT)
COMPARE
Waterproof Portable Meter, SJ All-in-One Probe, and Calibration
£668.32GBP / Each
£801.98GBP (incl VAT)
COMPARE
Waterproof Portable Meter; Meter Only with Calibration
£556.29GBP / Each
£667.55GBP (incl VAT)
COMPARE
Waterproof Portable Meter Kit
£714.00GBP / Each
£856.80GBP (incl VAT)
COMPARE
Waterproof Portable Meter Kit with Calibration
£780.86GBP / Each
£937.03GBP (incl VAT)