Monarch Instrument Nova-Strobe Digital Stroboscopes
Measure stop-motion quickly and easily
  • Microprocessor ensures measurement accuracy
These lightweight, digital stroboscopes make stop-motion measurements quickly and easily—just point, shoot, and adjust flash rate. Adjust the flash rate with digital encoder knob on the side. Last flash rate used is stored in memory, and will display on LCD when strobe is reactivated. For hands-free operation, activate trigger lock, and mount the stroboscope on a tripod.
Stroboscopes are available with AC power or rechargeable internal batteries, and as stroboscopes only or stroboscope kits.
£373.19 - £703.30GBP / Each
£447.83 - £843.96GBP (incl VAT)
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Display Type Power (VAC)
Availability Pricing
Monarch 6206-010 Nova-Strobe BAX Digital Stroboscope; 115 VAC WZ-87701-01 Mfr # 6206-010
COMPARE
6-digit backlit LCD
£373.19GBP / Each
£447.83GBP (incl VAT)
Monarch 6206-011 Nova-Strobe BAX Stroboscope Kit; 115V WZ-87701-02 Mfr # 6206-011
COMPARE
6-digit backlit LCD
£506.37GBP / Each
£607.64GBP (incl VAT)
Monarch 6206-012 Nova-Strobe BAX Digital Stroboscope; 230 VAC WZ-87701-03 Mfr # 6206-012
COMPARE
6-digit backlit LCD
£407.05GBP / Each
£488.46GBP (incl VAT)
Monarch 6206-013 Nova-Strobe BAX Stroboscope Kit; 230V WZ-87701-04 Mfr # 6206-013
COMPARE
6-digit backlit LCD
£543.19GBP / Each
£651.83GBP (incl VAT)
Monarch 6207-012 Nova Strobe bbx Stroboscope, battery powered, with 115/230 VAC recharger WZ-87701-05 Mfr # 6207-012
COMPARE
6-digit backlit LCD
£522.40GBP / Each
£626.88GBP (incl VAT)
Monarch 6207-013 6207-011 Nova-Strobe BBX Digital Stroboscope Kit WZ-87701-06 Mfr # 6207-013
COMPARE
6-digit backlit LCD
£703.30GBP / Each
£843.96GBP (incl VAT)