This product is now discontinued, View Related Products
Multipro Sector Probes
Ideal for sampling powders, grains, seeds, and pellets
  • Sector probe is made of anodized aluminum with stainless steel tip
  • Can be sterilized
This multi-sectored sampler is ideal for sampling powders, pellets, and other materials which may become non-homogeneous in bags at various levels. To collect sample, turn the T-shaped handle to close the cells and insert the steel point into the container. When the sampler is deep enough to cover all three sectors, turn the handle half-way to open the cells which allow the samples to enter. Then, return handle to its original position. The sample can now be removed and the different samples place into containers for analysis.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Diameter (in) Length (in)
Availability Pricing
COMPARE
0.75 21.75
Call For Price
COMPARE
0.75 31.5