Multifunctional Portable Indoor Air Quality Monitor
Portable monitor works on line power or rechargeable battery pack for location flexibility
  • Real-time data logging with download using USB or Bluetooth® wireless connectivity
  • Ideal for labs, cleanrooms, schools, and much more
  • Swivel handle for easy transport
Rechargeable battery pack, lightweight design, and swivel handle allows you to take this monitor wherever IAQ testing is needed. Built-in sampling pump continuously draws in an air sample for analysis. All models include sensors for carbon dioxide, ambient temperature, humidity, barometric pressure, with calculated values for dew point and wet bulb temperature.
Store up to 1000 sets of air quality data. Select a data set to manually store or set a logging interval from 10 seconds to 60 minutes for automatic data capture. Customize the name of a data set for easier record keeping and trend analysis.
Connect to a computer using the USB interface or wirelessly using Bluetooth® wireless connectivity. For computers without Bluetooth® wireless connectivity, a receiver is included that plugs into a USB port. The included software allows you to monitor, record min/max, retrieve stored data, set alarms, and plot graphs for analysis. The optional Bluetooth® printer runs on rechargeable batteries for easy portability.
Order the optional remote probe with 10-ft (3-m) hose for use in hard-to-reach areas. Probe is good for temperatures up to 1470°F (800°C) and includes a built-in thermcouple for temperature measurement.
£6,542.73GBP / Each
£7,851.28GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
E Instruments EXPERT-1-C1-V1 Portable Indoor Air Quality Monitor with Carbon Monoxide and VOC sensors WZ-81974-02 Mfr # EXPERT-1-C1-V1
COMPARE
Portable Indoor Air Quality Monitor with Carbon Monoxide and VOC sensors
£6,542.73GBP / Each
£7,851.28GBP (incl VAT)