This product is now discontinued, View Related Products
MultiDispo Disposable Solids Samplers
Easy zone sampling for analysis
  • Three openings sample different layers
  • Open-ended handle transfers sample to waiting container
  • Suitable for free-flowing powders and small-grain granulates
  • Gamma sterilized and nonsterile versions available
This HDPE solids sampler features three openings along the hollow shaft to take samples from three different zones, perfect for applications in which cross-sections are desired. The handle is open at the end, allowing you to transfer your sample to a container simply by tilting the sampler.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Length (in) Description
Availability Pricing
Dynalon MultiDispo Solids Sampler w/ Three Zones, 100 mL, 500 mm L; 20/pk WZ-33687-06 Mfr # 853933-4440
COMPARE
19.625 Solids Sampler w/ Three Zones, 100 mL, 500 mm L; 20/pk
Call For Price
Dynalon MultiDispo Sterile Solids Sampler w/ Three Zones, 100 mL, 500 mm L; 20/pk WZ-33687-07 Mfr # 853933-4441
COMPARE
39.375 Sterile Solids Sampler w/ Three Zones, 100 mL, 500 mm L; 20/pk
Call For Price
Dynalon MultiDispo Solids Sampler w/ Three Zones, 200 mL, 1 m L; 20/pk WZ-33687-08 Mfr # 853933-4450
COMPARE
19.625 Solids Sampler w/ Three Zones, 200 mL, 1 m L; 20/pk
Call For Price
Dynalon MultiDispo Sterile Solids Sampler w/ Three Zones, 200 mL, 1 m L; 20/pk WZ-33687-09 Mfr # 853933-4451
COMPARE
39.375 Sterile Solids Sampler w/ Three Zones, 200 mL, 1 m L; 20/pk