This product is now discontinued, View Related Products
Motorized Cap Torque Testers
Precision accuracy ensures quality control
  • Automatic cap torque testing for both tightening and untightening
  • Tighten caps with pre-programmed torque values
  • Accuracy of ±0.5% full scale, ±1 LSD
  • User selectable units of measure
  • Real time and peak mode (selectable)
  • 1,000 data points of memory
  • USB, RS-232C, and analog outputs
  • Capping speed 3 to 90° per second
  • Overload capacity 150% of full scale, with alarm at 110%full scale
  • Heavy duty all-metal construction for exceptional durability
These cap torque testers allow you to operate in both manual and automatic modes for additional flexibility. Manual mode is ideal for capturing peak torque applied to already tightened caps while the automatic mode allows you to apply pre-programmed torque values to caps. Units feature programmable high/low set points with LED indicators for easy go/no go testing. The yellow LED indicates measurements less than the low set point, the green LED indicates measurements that fall between high and low set points, and the red LED is activated for measurements that fall over the high set point and automatically stops testing.
Units operate in clockwise or counterclockwise directions and accommodate sample container diameters from .8" to 7", cap diameters from .8" to 3.5", and container heights from 3.2" to 9.4". All models include an extension shaft pin for short container applications.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Capacity (lb-in) Capacity (N-cm)
Availability Pricing
Imada MTG-15 Digital Cap Torque Tester, 15 lbf. WZ-25301-46 Mfr # MTG-15
COMPARE
15 200
Call For Price
Imada MTG-40 Digital Cap Torque Tester, 40.00 lbf. WZ-25301-47 Mfr # MTG-40
COMPARE
40 500
Call For Price
Imada MTG-85 Digital Cap Torque Tester, 85.0 lbf. WZ-25301-48 Mfr # MTG-85
COMPARE
85 1000