Millipore® Stirred Cells
Five different sizes handle volumes from 3 to 400 mL
  • Feature high flow rates with solutions up to 10% macrosolute concentration
Use these stirred cells to concentrate, diafilter, and exchange buffers for macromolecule solutions including proteins, enzymes, antibodies, and viruses. Maximum operating pressure is 75 psi. Recommended operating pressure is 55 psi (< 10 psi when using a 100K MWCO membrane or greater). Autoclavable at 121°C for 30 minutes.
£1,270.91GBP / Each
£1,525.09GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Membrane Diameter (mm) Description
Availability Pricing
EMD Millipore UFSC05001 5122 Amicon Stirred Cell Model 8050, 50 mL for 44.5 mm Filters; 1/Pk WZ-29968-14 Mfr # UFSC05001
COMPARE
44.5 5122 Amicon Stirred Cell Model 8050, 50 mL for 44.5 mm Filters; 1/Pk
£1,270.91GBP / Each
£1,525.09GBP (incl VAT)