Millipore® Millipak™ Filter Units
Pump-driven device for volumes from 10 to 30 L
  • Maximum inlet pressure is 75 psi at 25 °C
These filter units have a unique stacked-disk design to eliminate the particle-generating woven down-stream support found in pleated cartridges. They feature polycarbonate housing and hydrophilic PVDF filter membrane. The units have ¼" inlet and outlet hose barb connections. Supplied gamma sterilized and maybe autoclaved up to three times.
£278.40 - £347.11GBP / Pkg of 2
£334.08 - £416.53GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Membrane Material Pore Size (µm) Sample Volume (mL)
Availability Pricing
EMD Millipore MPGL02GH2 20 Filter, 10L, 0.22um PVDF, 1/4" ID, Sterile; 2/Pk WZ-29969-54 Mfr # MPGL02GH2
COMPARE
Polypropylene 0.22 100
£278.40GBP / Pkg of 2
£334.08GBP (incl VAT)
EMD Millipore MPGL06GH2 60 Filter, 30L, 0.22um PVDF, 1/4" ID, Sterile; 2/Pk WZ-29969-58 Mfr # MPGL06GH2
COMPARE
Polypropylene 0.22 300
£347.11GBP / Pkg of 2
£416.53GBP (incl VAT)