Micropump® Suction Shoe Gear Pumps for NEMA 56 C Motors
Easily attach your gear pump to a motor
  • Suction shoe design gives higher delivery pressures and superior priming performance.
  • Smooth, pulseless flow
  • Precision, leak-free delivery of fluids
  • Compact and easy to service
  • Use with any NEMA 56 C motors
Magnetically driven, precision-geared pump with unique suction shoe design provides smooth, accurate, pulseless fluid delivery. The magnetic drive, Viton® fluoroelastomer O-rings, and PTFE seal keep fluids securely inside and potential contaminants out. Chemically resistant to a wide range of materials. Modular design offers flexibility to configure to your specific needs.
£1,748.72 - £2,794.26GBP / Each
£2,098.46 - £3,353.11GBP (incl VAT)
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Differential Pressure (PSI) Flow Rate (mL/rev) Wetted Parts
Availability Pricing
Micropump NEMA 56C Suction Shoe Pump Head, 120psi/SS/PPS/Viton, 1.82 mL/rev WZ-07003-32 Mfr # 85719
COMPARE
120 1.82
£1,827.81GBP / Each
£2,193.37GBP (incl VAT)
Micropump NEMA 56C Suction Shoe Pump Head; SS/PPS/Viton; 3.48 mL/rev WZ-07003-33 Mfr # 81377 (GC-M35.PVS.E)
COMPARE
50 3.48
£2,001.16GBP / Each
£2,401.39GBP (incl VAT)
Micropump NEMA 56C Suction Shoe Pump Head; SS/PPS/Viton; 0.81 mL/rev WZ-07003-41 Mfr # 81946 (GC-M23.PVS.E)
COMPARE
75 0.81
£1,748.72GBP / Each
£2,098.46GBP (incl VAT)
Micropump Suction Shoe Gear Pump For NEMA 56 C Motors, 1.82 mL/rev, Peek Gears WZ-73005-02 Mfr # L18413
COMPARE
120 1.82
£1,844.06GBP / Each
£2,212.87GBP (incl VAT)
Micropump L20563 NEMA 56C Suction Shoe Pump Head; SS/PEEK/TEV; 0.81 mL/rev WZ-73005-05 Mfr # L18039
COMPARE
65 0.81
£2,332.70GBP / Each
£2,799.24GBP (incl VAT)
Micropump NEMA 56C Suction Shoe Pump Head, 75psi/SS/PPS/Viton, 1.82 mL/rev WZ-73005-07 Mfr # 81376
COMPARE
65 1.82
£1,953.49GBP / Each
£2,344.19GBP (incl VAT)
Micropump L20563 NEMA 56C Suction Shoe Pump Head; SS/PPS/EPDM; 1.82 mL/rev WZ-73005-08 Mfr # L19053
COMPARE
65 1.82
£2,017.41GBP / Each
£2,420.89GBP (incl VAT)
Micropump L20563 NEMA 56C Suction Shoe Pump Head; SS/PEEK/Viton; 1.82 mL/rev WZ-73005-09 Mfr # 210008
COMPARE
65 1.82
£2,079.17GBP / Each
£2,495.00GBP (incl VAT)
Micropump L20563 NEMA 56C Suction Shoe Pump Head; SS/PEEK/TEV; 3.48 mL/rev WZ-73005-13 Mfr # L18542
COMPARE
65 3.48
£2,794.26GBP / Each
£3,353.11GBP (incl VAT)
Showing 1- 9 of 9