Micropump Cavity-Style Gear Pump Heads for NEMA Type 56 C-face Motors
Smooth pulseless flow provides precise, leak-free delivery of fluids
  • Compact size and easy to service
  • Cavity-style heads are designed to handle higher inlet pressures.
Magnetically driven, precision-geared cavity pump heads provide smooth, accurate, pulseless fluid delivery. The magnetic drive and a single seal keep fluids securely inside and potential contaminants out. Chemical resistant materials handle a wide range of fluids.
£2,567.64 - £6,384.87GBP / Each
£3,081.17 - £7,661.84GBP (incl VAT)
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Differential Pressure (PSI) Flow Rate (mL/rev) Max Temperature (° F)
Availability Pricing
Micropump L20563 NEMA 56C Cavity Style Pump Head; 60psi/SS/PEEK/TEV; 3.48 mL/rev WZ-73007-00 Mfr # L20563
COMPARE
60 3.48 200
£2,567.64GBP / Each
£3,081.17GBP (incl VAT)
Micropump NEMA 56C Cavity Style Pump Head; 100psi/SS/PEEK/TEV; 3.48 mL/rev WZ-73007-02 Mfr # L26621
COMPARE
100 3.48 200
£3,172.39GBP / Each
£3,806.87GBP (incl VAT)
Micropump 200037 NEMA 56C Cavity Style Pump Head; 50psi/SS/PEEK/PTFE; 7.7 mL/rev WZ-73007-11 Mfr # 200037
COMPARE
50 7.7 200
£6,384.87GBP / Each
£7,661.84GBP (incl VAT)