Micropump B-Mount Gear Pump Heads
Smooth pulseless flow provides precise, leak-free delivery of fluids
  • Suction shoe designs provides higher delivery pressures and temperatures, plus increased efficiency
  • Cavity style designs with no dynamic seals for applications that cannot tolerate leaks
Magnetically driven, precision-geared pump heads provide smooth, accurate, pulseless fluid delivery. The magnetic drive and static seals keep fluids securely inside and potential contaminants out. Chemical resistant materials handle a wide range of materials (due to wetted parts). Pump heads are compact and easy to service. Modular design offers flexibility—configure to your specific needs.
£820.56 - £1,810.19GBP / Each
£984.67 - £2,172.23GBP (incl VAT)
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Flow Rate (mL/rev) Gear Material Seal Material
Availability Pricing
Micropump GJ Series B-Mount Cavity Gear Pump Head, SS Base, PTFE Gear, PTFE Seal, Internal Bypass; 0.64 mL/rev SKU 7400401 WZ-74004-01 Mfr # MI0013 / Z-120 / 82004
COMPARE
0.64 PTFE PTFE
£882.49GBP / Each
£1,058.99GBP (incl VAT)
Micropump GJ Series B-Mount Cavity Gear Pump Head, SS Base, PTFE Gear, PTFE Seal, Internal Bypass; 0.91 mL/rev WZ-74004-03 Mfr # MI0015 / Z-122 / 82006
COMPARE
0.91 PTFE PTFE
£882.49GBP / Each
£1,058.99GBP (incl VAT)
Micropump GJ Series B-Mount Cavity Gear Pump Head, SS Base, PPS Gear, PTFE Seal, Internal Bypass; 0.64 mL/rev WZ-74004-05 Mfr # MI0019 / Z-130 / 81529
COMPARE
0.64 PPS PTFE
£882.28GBP / Each
£1,058.74GBP (incl VAT)
Micropump GJ Series B-Mount Cavity Gear Pump Head, SS Base, PTFE Gear, PTFE Seal; 0.316 mL/rev WZ-74004-07 Mfr # MI0267 / Z-040 / 81977
COMPARE
0.316 PTFE PTFE
£820.56GBP / Each
£984.67GBP (incl VAT)
Micropump GJ Series B-Mount Cavity Gear Pump Head, SS Base, PTFE Gear, PTFE Seal; 0.64 mL/rev WZ-74004-09 Mfr # MI0016 / Z-140 / 82001
COMPARE
0.64 PTFE PTFE
£833.24GBP / Each
£999.89GBP (incl VAT)
Micropump GJ Series B-Mount Cavity Gear Pump Head, SS Base, PTFE Gear, PTFE Seal; 0.91 mL/rev WZ-74004-11 Mfr # MI0018 / Z-142 / 82003
COMPARE
0.91 PTFE PTFE
£837.77GBP / Each
£1,005.32GBP (incl VAT)
Micropump GJ Series B-Mount Cavity Gear Pump Head, SS Base, PPS Gear, PTFE Seal; 0.64 mL/rev WZ-74004-17 Mfr # MI0020 / Z-150 / 81531
COMPARE
0.64 PPS PTFE
£837.56GBP / Each
£1,005.07GBP (incl VAT)
Micropump GA Series B-Mount Suction Shoe Gear Pump Head, SS Base, Graphite Gear, PTFE Seal; 0.017 mL/rev WZ-74004-23 Mfr # MI0006 / Z-186 / 82092
COMPARE
0.017 Carbon graphite PTFE
£931.96GBP / Each
£1,118.35GBP (incl VAT)
Micropump GA Series B-Mount Suction Shoe Gear Pump Head, SS Base, PPS Gear, PTFE Seal; 0.017 mL/rev WZ-74004-27 Mfr # MI0278 / Z-186 PPS / L18412
COMPARE
0.017 PPS PTFE
£933.21GBP / Each
£1,119.85GBP (incl VAT)
Micropump GA Series B-Mount Suction Shoe Gear Pump Head, SS Base, Graphite Gear, PTFE Seal; 0.042 mL/rev WZ-74004-29 Mfr # MI0007 / Z-181 / 82114
COMPARE
0.042 Carbon graphite PTFE
£978.21GBP / Each
£1,173.85GBP (incl VAT)
Micropump GA Series B-Mount Suction Shoe Gear Pump Head, SS Base, Graphite Gear, PTFE Seal; 0.084 mL/rev WZ-74004-31 Mfr # MI0008 / Z-183 / 82115
COMPARE
0.084 Carbon graphite PTFE
£978.21GBP / Each
£1,173.85GBP (incl VAT)
Micropump GJ Series B-Mount Cavity Gear Pump Head, SS Base, PTFE Gear, PTFE Seal, Internal Bypass; 0.64 mL/rev SKU 7400433 WZ-74004-33 Mfr # MI0131 / Z-1830 / 82004
COMPARE
0.64 PTFE PTFE
£880.40GBP / Each
£1,056.48GBP (incl VAT)
Micropump GA Series B-Mount Suction Shoe Gear Pump Head, SS Base, PEEK Gear, PTFE Seal; 0.017 mL/rev WZ-74004-71 Mfr # MI0312 / Z-186P / L20820
COMPARE
0.017 PEEK PTFE
£1,810.19GBP / Each
£2,172.23GBP (incl VAT)
Micropump GA Series B-Mount Suction Shoe Gear Pump Head, SS Base, PEEK Gear, PTFE Seal; 0.092 mL/rev WZ-74004-73 Mfr # MI0280 / Z-1830P / L18489
COMPARE
0.092 PEEK PTFE
£964.89GBP / Each
£1,157.87GBP (incl VAT)
Micropump GJ Series B-Mount Cavity Gear Pump Head, SS Base, PEEK Gear, PTFE Seal; 0.64 mL/rev WZ-74004-75 Mfr # MI0313 / Z-140P / L19735
COMPARE
0.64 PEEK PTFE
£958.64GBP / Each
£1,150.37GBP (incl VAT)
Showing 1- 15 of 15