Micropump® A-Mount Suction Shoe Gear Pump Heads
Smooth pulseless flow provides precise, leak-free delivery of fluids
  • Suction shoe design provides higher delivery pressures and temperatures, plus increased efficiency.
Magnetically driven, precision-geared pump heads provide smooth, accurate, pulseless fluid delivery. The magnetic drive and static seals keep fluids securely inside and potential contaminants out. Chemical resistant materials handle a wide range of materials (due to wetted parts). Pump heads are compact and easy to service. Modular design offers flexibility—configure to your specific needs.
£712.62 - £1,153.86GBP / Each
£855.14 - £1,384.63GBP (incl VAT)
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Flow Rate (mL/rev) Description
Availability Pricing
Micropump L20013 A-Mount Suction Shoe Pump Head; SS/PPS/PTFE; 0.092 mL/rev WZ-07002-14 Mfr # L20013
COMPARE
0.092 A-Mount Suction Shoe Pump Head; SS/PPS/PTFE; 0.092 mL/rev
£1,106.40GBP / Each
£1,327.68GBP (incl VAT)
Micropump L21834 A-Mount Suction Shoe Pump Head; SS/Graphite/PTFE; 0.017 mL/rev WZ-07002-25 Mfr # L21834
COMPARE
0.017 A-Mount Suction Shoe Pump Head; SS/Graphite/PTFE; 0.017 mL/rev
£712.62GBP / Each
£855.14GBP (incl VAT)
Micropump L21835 A-Mount Suction Shoe Pump Head; SS/Graphite/PTFE; 0.042 mL/rev WZ-07002-26 Mfr # L21835
COMPARE
0.042 A-Mount Suction Shoe Pump Head; SS/Graphite/PTFE; 0.042 mL/rev
£716.90GBP / Each
£860.28GBP (incl VAT)
Micropump L21836 A-Mount Suction Shoe Pump Head; SS/Graphite/PTFE; 0.084 mL/rev WZ-07002-27 Mfr # L21836
COMPARE
0.084 A-Mount Suction Shoe Pump Head; SS/Graphite/PTFE; 0.084 mL/rev
£729.74GBP / Each
£875.69GBP (incl VAT)
Micropump L22063 A-Mount Suction Shoe Pump Head; Bypass/SS/PPS/Viton; 0.58 mL/rev WZ-07003-02 Mfr # 85716
COMPARE
0.58 A-Mount Suction Shoe Pump Head; Bypass/SS/PPS/Viton; 0.58 mL/rev
£1,012.02GBP / Each
£1,214.42GBP (incl VAT)
Micropump L22094 A-Mount Suction Shoe Pump Head; Bypass/SS/PPS/Viton; 1.17 mL/rev WZ-07003-04 Mfr # 85717
COMPARE
1.17 A-Mount Suction Shoe Pump Head; Bypass/SS/PPS/Viton; 1.17 mL/rev
£1,153.86GBP / Each
£1,384.63GBP (incl VAT)
Micropump L22097 A-Mount Suction Shoe Pump Head; SS/PEEK/PTFE; 0.092 mL/rev WZ-73003-14 Mfr # L22097
COMPARE
0.092 A-Mount Suction Shoe Pump Head; SS/PEEK/PTFE; 0.092 mL/rev
£747.93GBP / Each
£897.52GBP (incl VAT)
Micropump L20561 A-Mount Suction Shoe Pump Head; Bypass/SS/PEEK/Viton; 0.261 mL/rev WZ-73004-00 Mfr # 85713
COMPARE
0.261 A-Mount Suction Shoe Pump Head; Bypass/SS/PEEK/Viton; 0.261 mL/rev
£741.51GBP / Each
£889.81GBP (incl VAT)
Micropump L3468 A-Mount Suction Shoe Pump Head; Bypass/SS/PEEK/Viton; 0.56 mL/rev WZ-73004-02 Mfr # 85714
COMPARE
0.58 A-Mount Suction Shoe Pump Head; Bypass/SS/PEEK/Viton; 0.56 mL/rev
£745.79GBP / Each
£894.95GBP (incl VAT)
Micropump L20562 A-Mount Suction Shoe Pump Head; Bypass/SS/PEEK/Viton; 1.12 mL/rev WZ-73004-04 Mfr # 85715
COMPARE
1.17 A-Mount Suction Shoe Pump Head; Bypass/SS/PEEK/Viton; 1.12 mL/rev
£867.77GBP / Each
£1,041.32GBP (incl VAT)
Micropump GB Series A-Mount Suction Shoe Pump Head, Bypass/SS/PPS/Viton; 0.58 mL/rev WZ-74004-19 Mfr # MI0022 / ZP-200 / 81281
COMPARE
Gear Pump Head, Suction Shoe Style, Stainless Steel, PPS Gear, Viton Seal, 0.58 mL/rev
£898.87GBP / Each
£1,078.64GBP (incl VAT)
Micropump GB Series A-Mount Suction Shoe Pump Head, Bypass/SS/PPS/Viton; 1.17 mL/rev WZ-74004-21 Mfr # MI0023 / ZP-201 / 81282
COMPARE
Gear Pump Head, Suction Shoe Style, Stainless Steel, PPS Gear, Viton Seal, 1.17 mL/rev
£1,048.08GBP / Each
£1,257.70GBP (incl VAT)
Micropump GB Series A-Mount Suction Shoe Pump Head, SS/PEEK/Viton; 0.58 mL/rev WZ-74004-77 Mfr # MI0306 / Z-200P / 220004
COMPARE
Gear Pump Head, Suction Shoe Style, Stainless Steel, PEEK Gear, Viton Seal, 0.58 mL/rev
£972.11GBP / Each
£1,166.53GBP (incl VAT)
Showing 1- 13 of 13