Microflex Cleanroom Gloves
Low surface particulates, ionic extractables, and non-volatile residues
Cleanroom gloves are washed in a microprocessor-controlled washer/dryer system to insure low residues. Micro-textured surface provides wet or dry grip. Disposable; for single use only. Ambidextrous.
Compliance: ISO Class 4 or 5 (see table).
£20.80 - £30.76GBP / Bag of 100
£24.96 - £36.91GBP (incl VAT)
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compliance Material Size
Availability Pricing
Microflex Latex Cleanroom Glove, ISO Class 5 compliant, small, 100/bag WZ-86313-15 Mfr # CE5-512-S
COMPARE
ISO Class 5 Latex Small
£23.89GBP / Bag of 100
£28.67GBP (incl VAT)
Microflex Latex Cleanroom Glove, ISO Class 5 compliant, medium, 100/bag WZ-86313-16 Mfr # CE5-512-M
COMPARE
ISO Class 5 Latex Medium
£23.89GBP / Bag of 100
£28.67GBP (incl VAT)
Microflex Latex Cleanroom Glove, ISO Class 5 compliant, large, 100/bag WZ-86313-17 Mfr # CE5-512-L
COMPARE
ISO Class 5 Latex Large
£23.89GBP / Bag of 100
£28.67GBP (incl VAT)
Microflex Latex Cleanroom Glove, ISO Class 5 compliant, extra large, 100/bag WZ-86313-18 Mfr # CE5-512-XL
COMPARE
ISO Class 5 Latex X-Large
£23.89GBP / Bag of 100
£28.67GBP (incl VAT)
Microflex Nitrile Cleanroom Glove, ISO Class 5 compliant, small, 100/bag WZ-86313-25 Mfr # CE5-755-S
COMPARE
ISO Class 5 Nitrile Small
£26.21GBP / Bag of 100
£31.45GBP (incl VAT)
Microflex Nitrile Cleanroom Glove, ISO Class 5 compliant, medium, 100/bag WZ-86313-26 Mfr # CE5-755-M
COMPARE
ISO Class 5 Nitrile Medium
£20.80GBP / Bag of 100
£24.96GBP (incl VAT)
Microflex Nitrile Cleanroom Glove, ISO Class 5 compliant, large, 100/bag WZ-86313-27 Mfr # CE5-755-L
COMPARE
ISO Class 5 Nitrile Large
£20.80GBP / Bag of 100
£24.96GBP (incl VAT)
Microflex Nitrile Cleanroom Glove, ISO Class 5 compliant, extra large, 100/bag WZ-86313-28 Mfr # CE5-755-XL
COMPARE
ISO Class 5 Nitrile X-Large
£26.21GBP / Bag of 100
£31.45GBP (incl VAT)
Microflex Latex Cleanroom Glove, ISO Class 4 compliant, small, 100/bag WZ-86313-35 Mfr # CE4-200-S
COMPARE
ISO Class 4 Latex Small
£28.08GBP / Bag of 100
£33.70GBP (incl VAT)
Microflex Latex Cleanroom Glove, ISO Class 4 compliant, medium, 100/bag WZ-86313-36 Mfr # CE4-200-M
COMPARE
ISO Class 4 Latex Medium
£28.08GBP / Bag of 100
£33.70GBP (incl VAT)
Microflex Latex Cleanroom Glove, ISO Class 4 compliant, large, 100/bag WZ-86313-37 Mfr # CE4-200-L
COMPARE
ISO Class 4 Latex Large
£28.08GBP / Bag of 100
£33.70GBP (incl VAT)
Microflex Latex Cleanroom Glove, ISO Class 4 compliant, extra large, 100/bag WZ-86313-38 Mfr # CE4-200-XL
COMPARE
ISO Class 4 Latex X-Large
£28.08GBP / Bag of 100
£33.70GBP (incl VAT)
Microflex Nitrile Cleanroom Glove, ISO Class 4 compliant, small, 100/bag WZ-86313-45 Mfr # CE4-313-S
COMPARE
ISO Class 4 Nitrile Small
£30.76GBP / Bag of 100
£36.91GBP (incl VAT)
Microflex Nitrile Cleanroom Glove, ISO Class 4 compliant, medium, 100/bag WZ-86313-46 Mfr # CE4-313-M
COMPARE
ISO Class 4 Nitrile Medium
£30.76GBP / Bag of 100
£36.91GBP (incl VAT)
Microflex Nitrile Cleanroom Glove, ISO Class 4 compliant, large, 100/bag WZ-86313-47 Mfr # CE4-313-L
COMPARE
ISO Class 4 Nitrile Large
£30.76GBP / Bag of 100
£36.91GBP (incl VAT)
Microflex Nitrile Cleanroom Glove, ISO Class 4 compliant, extra large, 100/bag WZ-86313-48 Mfr # CE4-313-XL
COMPARE
ISO Class 4 Nitrile X-Large
£30.76GBP / Bag of 100
£36.91GBP (incl VAT)
Showing 1- 16 of 16