This product is now discontinued, View Related Products
MicroDispo Disposable Small-Scale Solids Samplers
Don't waste valuable product
  • Tee-shaped handle opens and closes tip for precise sampling locations and small sample sizes
  • Free-flowing powders and small-grain bulk materials work best
  • Gamma sterilized and nonsterile versions available
This HDPE solids sampler features a tip that opens to allow sample to enter and then closes to secure the sample and ensure the correct location was sampled. The ergonomic tee-shaped handle operates the mechanism to open and close the tip with a push/pull action. Sample sizes as small as 10 mL are possible.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Length (in) Description
Availability Pricing
Dynalon MicroDispo Small Scale Solids Sampler, 10 mL, 500 mm L; 20/pk WZ-33687-51 Mfr # 853933-2240
COMPARE
19.625 Small Scale Solids Sampler, 10 mL, 500 mm L; 20/pk
Call For Price
Dynalon MicroDispo Sterile Small Scale Solids Sampler, 10 mL, 500 mm L; 20/pk WZ-33687-53 Mfr # 853933-2241
COMPARE
19.625 Sterile Small Scale Solids Sampler, 10 mL, 500 mm L; 20/pk
Call For Price
Dynalon MicroDispo Small Scale Solids Sampler, 10 mL, 1 m L; 20/pk WZ-33687-55 Mfr # 853933-2250
COMPARE
39.375 Small Scale Solids Sampler, 10 mL, 1 m L; 20/pk
Call For Price
Dynalon MicroDispo Sterile Small Scale Solids Sampler, 10 mL, 1 m L; 20/pk WZ-33687-57 Mfr # 853933-2251
COMPARE
39.375 Sterile Small Scale Solids Sampler, 10 mL, 1 m L; 20/pk