METTLER TOLEDO SevenExcellence™ Benchtop pH/mV Meters
Improve efficiency and productivity
  • Intuitive plain-language interface and touch screen allow rapid familiarization
  • Predefined and custom methods provide the results you need without the hassle of setup
  • PIN-protected user management functions provide security for compliance with regulatory agencies
These pH meters feature an interface designed to improve work flow. The large touch screen makes navigation easy and the menu structure uses plain language, eliminating confusion during set up. One click will start one of 17 predefined or 60 custom methods, enhancing efficiency and throughput. Select the display that works best for you—uFocus™ shows only the most important data while the standard view shows more detail including setup parameters and measurements.
Security features help you comply with regulatory agency requirements. User management settings and PIN-protected functions prevent tampering with custom methods. Integrated Sensor Management (ISM®) pH electrodes are automatically recognized by the meter and store their own calibration data ensuring every measurement is accurate even if you switch electrodes. Time and date stamps from the radio-controlled clock ensure measurements conform to GLP guidelines.
Included electrode arm smoothly lowers and lifts the electrode vertically with one hand, reducing spills and time between measurements. Modular construction allows you to expand your measurement capabilities as your lab needs evolve. Connect with PCs and peripheral devices using the USB, Ethernet, and RS-232 interfaces.
£2,169.43 - £2,546.91GBP / Each
£2,603.32 - £3,056.29GBP (incl VAT)
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Mettler Toledo SevenExcellence S400 Benchtop pH/mV Meter Only WZ-41000-01 Mfr # 30046240
COMPARE
Benchtop pH/mV Meter Only
£2,169.43GBP / Each
£2,603.32GBP (incl VAT)
Mettler Toledo SevenExcellence S400 Benchtop pH/mV Meter Standard Kit WZ-41000-02 Mfr # 30046241
COMPARE
Benchtop pH/mV Meter Standard Kit
£2,434.10GBP / Each
£2,920.92GBP (incl VAT)
Mettler Toledo SevenExcellence S400 Benchtop pH/mV Meter Bio Kit WZ-41000-03 Mfr # 30046242
COMPARE
Benchtop pH/mV Meter Bio Kit
£2,546.91GBP / Each
£3,056.29GBP (incl VAT)
Mettler Toledo SevenExcellence S400 Benchtop pH/mV Meter Standard Kit with Magnetic Stirrer WZ-41000-05 Mfr # 30092983
COMPARE
Benchtop pH/mV Meter Standard Kit with Magnetic Stirrer
£2,546.91GBP / Each
£3,056.29GBP (incl VAT)