This product is now discontinued, View Related Products
METTLER TOLEDO SevenCompact™ S220 Benchtop pH/Ion Meter
Easily switch between pH and ion concentration measurements
  • Preprogrammed with the most common ion selective electrode types for convenience
  • A single key press toggles between pH and ion measurement modes
  • ISM® (Intelligent Sensor Management) pH and ion selective electrodes ensure data integrity
This benchtop pH and ion meter increases efficiency with one-touch operation. One key press begins the measurement, waits for stability, stores the stable reading with GLP stamp, and can automatically transfer the data to USB flash drive (not included). The user-friendly interface allows you to quickly and easily set up parameters, toggle between screens and functions, and change electrodes to keep pace with a busy lab.
Using the Intelligent Sensor Management (ISM) electrodes provides security and peace of mind. With automatic sensor identification and calibration data stored in the electrode's memory, you'll know you're getting accurate measurements with the correct sensor.
Durable construction with IP54 water and dust resistance, rubber caps for connectors, and transparent in-use cover protect the meter from splashes and spills.
Call For Price
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Mettler Toledo SevenCompact S220 Benchtop pH/Ion Meter Only WZ-41000-29 Mfr # 30019028
COMPARE
Benchtop pH/Ion Meter Only
Call For Price
Mettler Toledo SevenCompact S221 Benchtop pH/Ion Meter Standard Kit WZ-41000-30 Mfr # 30019029
COMPARE
Benchtop pH/Ion Meter Standard Kit
Call For Price
Mettler Toledo SevenCompact S222 Benchtop pH/Ion Meter Bio Kit WZ-41000-31 Mfr # 30019031
COMPARE
Benchtop pH/Ion Meter Bio Kit
Call For Price
Mettler Toledo SevenCompact S220 Benchtop pH/Ion Meter Universal Kit WZ-41000-32 Mfr # 30019032
COMPARE
Benchtop pH/Ion Meter Universal Kit
Call For Price
Mettler Toledo SevenCompact S220 Micro Kit Benchtop pH/Ion Meter Micro Kit WZ-41000-33 Mfr # 30096985
COMPARE
Benchtop pH/Ion Meter Micro Kit
Call For Price
Mettler Toledo SevenCompact S220 uMix Kit Benchtop pH/Ion Meter Standard Kit with Magnetic Stirrer WZ-41000-34 Mfr # 30096986
COMPARE
Benchtop pH/Ion Meter Standard Kit with Magnetic Stirrer