This product is now discontinued, View Related Products
METTLER TOLEDO S230 SevenCompact Benchtop Conductivity Meters
Boost productivity—automation saves time
  • A single touch measures, stores, and transfers readings leaving you free to perform other tasks
  • Intuitive plain-language menu is easy to navigate and set up measurement parameters
  • Transfer saved data to a USB flash drive and download to PC for analysis
This benchtop conductivity meter increases efficiency with one-touch operation. One key press begins the measurement, waits for stability, stores the stable reading with GLP stamp, and can automatically transfer the data to USB flash drive (not included). The user-friendly interface allows you to quickly and easily set up parameters, toggle between screens and functions, and change cells to keep pace with a busy lab.
Using the Intelligent Sensor Management (ISM) cells provides security and peace of mind. With automatic sensor identification and calibration data stored in the cell's memory, you'll know you're getting accurate measurements with the correct sensor.
Durable construction with IP54 water and dust resistance, rubber caps for connectors, and transparent in-use cover protect the meter from splashes and spills.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Mettler Toledo S230 Benchtop Conductivity Meter Only WZ-41000-35 Mfr # 30019033
COMPARE
Benchtop Conductivity Meter Only
Call For Price
Mettler Toledo S230 Benchtop Conductivity Meter Standard Kit WZ-41000-36 Mfr # 30019034
COMPARE
Benchtop Conductivity Meter Standard Kit
Call For Price
Mettler Toledo S230 Benchtop Conductivity Meter USP/EP Kit WZ-41000-37 Mfr # 30019035
COMPARE
Benchtop Conductivity Meter USP/EP Kit