Mettler Toledo NewClassic ME-T Precision Toploading Legal for Trade (NTEP) Balances
Worry-free weighing with NTEP certification
  • Large color touchscreen and intuitive interface simplify operation
  • Fully Automatic Calibration Technology (FACT) ensures accuracy
  • Eight embedded applications include essential weighing functions, statistics, and formulation
  • Sample ID and passcode protection produce GLP documentation of results
  • RS-232 and USB connectivity simplify data upload and transfer
Precision toploading NTEP balances feature a 4.5” color TFT touchscreen with intuitive guidance enabling you to operate quickly and efficiently, even while wearing gloves. An electromagnetic force compensation (EMFC) weighing cell with fully automated calibration technology (FACT) automatically adjusts the balance to provide consistent reliable results. Multiple interfaces support barcode reader, printer and PC connections for easy traceability and data handling. USB and RS-232 connectivity along with an optional Bluetooth adapter (sold separately) simplify data upload and transfer.
The NTEP certification provides confidence that a device will be manufactured in accordance with United States standards.
£2,459.01 - £3,052.57GBP / Each
£2,950.81 - £3,663.08GBP (incl VAT)
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Readability (g)
Availability Pricing
Mettler Toledo NewClassic ME-T Legal for Trade (NTEP) Precision Toploading Balance, 320 g x 1 mg, Internal Calibration WZ-11348-16 Mfr # ME303T/A
COMPARE
Internal 320 0.001
£2,789.26GBP / Each
£3,347.11GBP (incl VAT)
Mettler Toledo NewClassic ME-T Legal for Trade (NTEP) Precision Toploading Balance, 1200 g x 10 mg, Internal Calibration WZ-11348-17 Mfr # ME1002T/A
COMPARE
Internal 1200 0.01
£2,459.01GBP / Each
£2,950.81GBP (incl VAT)
Mettler Toledo NewClassic ME-T Legal for Trade (NTEP) Precision Toploading Balance, 2200 g x 10 mg, Internal Calibration WZ-11348-18 Mfr # ME2002T/A
COMPARE
Internal 2200 0.01
£2,682.15GBP / Each
£3,218.58GBP (incl VAT)
Mettler Toledo NewClassic ME-T Legal for Trade (NTEP) Precision Toploading Balance, 3200 g x 10 mg, Internal Calibration WZ-11348-19 Mfr # ME3002T/A
COMPARE
Internal 3200 0.01
£2,882.98GBP / Each
£3,459.58GBP (incl VAT)
Mettler Toledo NewClassic ME-T Legal for Trade (NTEP) Precision Toploading Balance, 4200 g x 10 mg, Internal Calibration WZ-11348-20 Mfr # ME4002T/A
COMPARE
Internal 4200 0.01
£3,052.57GBP / Each
£3,663.08GBP (incl VAT)