Cole-Parmer® Mesh Insert Trays for Ultrasonic Cleaners
Achieve optimal cleaning results
  • Supports items in tank for direct cleaning
  • Prevents damage to transducers
Made of 304 stainless steel, these mesh trays can be used to support items in the tank that require direct cleaning. Ultrasonic activity is greatest about an inch from the bottom of the tank, so suspending your items provides the best cleaning possible and helps prevent damage to the transducers.
£88.53 - £140.89GBP / Each
£106.24 - £169.07GBP (incl VAT)
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Mesh tray for 1/2 Gallon and Stainless Steel 3/4 Gallon Cleaners.
£88.53GBP / Each
£106.24GBP (incl VAT)
COMPARE
Mesh tray for 3/4 Gallon and Stainless Steel 1 Gallon Cleaners.
£115.99GBP / Each
£139.19GBP (incl VAT)
COMPARE
Mesh Tray for 1.5 gal and SS 1.5 gal Cleaners
£140.89GBP / Each
£169.07GBP (incl VAT)