Megger® DET2/3 Earth Ground Tester
Increase safety and performance of electrical systems
  • Obtain measurements in 4P, resistivity, 3P, ART (selective), 2P, and stakeless test modes
  • Operate in the field all day—battery lasts long and recharges quickly
  • Collect high-accuracy earth measurements with 1 mΩ resolution
  • Withstand the elements—IP54 weatherproof with case open, IP65 waterproof when closed
  • Automatic resistance measurements from 0.01 to 200 Ω
This high-performance DET2/3 earth/ground meter provides step-by-step guidance to complete earth tests as required by published standards. Data points are recorded—up to 500 readings—a test report is then built and can be downloaded through USB to a PC. The instrument's battery can last up to 10 hours, and recharges to 100% in three hours—or operate from a standard 12 VDC vehicle power supply.
Order model 20000-96 to obtain wireless remote operation. This tester has additional features to record GPS coordinates, ambient temperature and humidity measurements, and wireless remote control for added safety.
£5,334.00 - £6,072.00GBP / Each
£6,400.80 - £7,286.40GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Resistance (k ohms) Description
Availability Pricing
Megger DET2/3 High Performance Ground Tester WZ-20000-95 Mfr # DET2/3
COMPARE
£5,334.00GBP / Each
£6,400.80GBP (incl VAT)
Megger DET2/3RC High Performance Ground Tester with Remote Control and GPS WZ-20000-96 Mfr # DET2/3RC
COMPARE
£6,072.00GBP / Each
£7,286.40GBP (incl VAT)