Masterflex® Variable Area Flowmeter Multitube Frames
Create your own flowmeter systems for complete flexibility
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate
  • Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate
  • Use with media for laboratory and industrial applications
  • Required system components for flowmeter systems would be frame, flowtube(s), valve cartridge, and tripod—order each component separately
  • Choose from aluminum or stainless steel wetted parts based on required chemical compatibility
  • Designed with 1/8” NPT(F) connections
Design a flow system to meet your metering, mixing, or monitoring applications. We offer a variety of frames and flowtubes so that you can create the system to meet your exact needs.
Choose a frame depending on the flowtube capacity, flow pattern, and wetted parts that you require.
These frames have valve ports which accept valve cartridges to control flow. A tripod base is required to use the multitube frame with the common inlet flow pattern; all other multitube frames are designed for panel and tripod mounting.
£99.68 - £881.94GBP / Each
£119.62 - £1,058.33GBP (incl VAT)
37 variations of this product are available.
See All Product Options Below
37 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Two Tube Frame for 65 mm Flowtube, Common, Aluminum
£123.52GBP / Each
£148.22GBP (incl VAT)
COMPARE
Two Tube Frame for 65 mm Flowtube, Common, 316 SS
£277.37GBP / Each
£332.84GBP (incl VAT)
COMPARE
Three Tube Frame for 65 mm Flowtube, Common, 316 SS
£398.72GBP / Each
£478.46GBP (incl VAT)
COMPARE
Four Tube Frame for 65 mm Flowtube, Common, Aluminum
£192.86GBP / Each
£231.43GBP (incl VAT)
COMPARE
Four Tube Frame for 65 mm Flowtube, Common, 316 SS
£522.23GBP / Each
£626.68GBP (incl VAT)
COMPARE
Five Tube Frame for 65 mm Flowtube, Isolated, Aluminum
£202.61GBP / Each
£243.13GBP (incl VAT)
COMPARE
Five Tube Frame for 65 mm Flowtube, Common, Aluminum
£236.20GBP / Each
£283.44GBP (incl VAT)
COMPARE
Five Tube Frame for 65 mm Flowtube, Isolated, 316 SS
£601.32GBP / Each
£721.58GBP (incl VAT)
COMPARE
Six Tube Frame for 65 mm Flowtube, Isolated, Aluminum
£231.86GBP / Each
£278.23GBP (incl VAT)
COMPARE
Six Tube Frame for 65 mm Flowtube, Common, 316 SS
£767.09GBP / Each
£920.51GBP (incl VAT)
COMPARE
Two Tube Frame for 150 mm Flowtube, Isolated, Aluminum
£140.85GBP / Each
£169.02GBP (incl VAT)
COMPARE
Two Tube Frame for 150 mm Flowtube, Common, 316 SS
£313.12GBP / Each
£375.74GBP (incl VAT)
COMPARE
Three Tube Frame for 150 mm Flowtube, Isolated, Aluminum
£183.11GBP / Each
£219.73GBP (incl VAT)
COMPARE
Three Tube Frame for 150 mm Flowtube, Common, Aluminum
£212.36GBP / Each
£254.83GBP (incl VAT)
COMPARE
Four Tube Frame for 150 mm Flowtube, Common, Aluminum
£269.78GBP / Each
£323.74GBP (incl VAT)
COMPARE
Four Tube Frame for 150 mm Flowtube, Common, 316 SS
£595.91GBP / Each
£715.09GBP (incl VAT)
COMPARE
Five Tube Frame for 150 mm Flowtube, Common, Aluminum
£315.29GBP / Each
£378.35GBP (incl VAT)
COMPARE
Five Tube Frame for 150 mm Flowtube, Common, 316 SS
£735.67GBP / Each
£882.80GBP (incl VAT)
COMPARE
Six Tube Frame for 150 mm Flowtube, Isolated, Aluminum
£342.38GBP / Each
£410.86GBP (incl VAT)
COMPARE
Six Tube Frame for 150 mm Flowtube, Common, Aluminum
£390.05GBP / Each
£468.06GBP (incl VAT)
Showing 1 - 20 of 37