Masterflex® Union Fittings, Sanitary Clamp, 45° Elbow, 316L Stainless Steel
Meets FDA requirements and 3A sanitary standards
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Easily connects pipes or fittings of different types
  • Compatible with standard 304L fitting clamps
  • Ideal for pharmaceutical industry
These 45-degree elbow adapters allow for secure connection of sanitary lines when a 45-degree angle is required. The 316L stainless steel couplings are suitable for the pharmaceutical industry. They offer great chemical compatibility for use with corrosive acids and bases. Clamps and gaskets required for a complete system are sold separately.
£47.67 - £102.93GBP / Each
£57.20 - £123.52GBP (incl VAT)
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Fitting Material Sanitary Clamp Size (Port 1) (in) Sanitary Clamp Size (Port 2) (in)
Availability Pricing
COMPARE
316L Stainless Steel 3/4 3/4
£102.93GBP / Each
£123.52GBP (incl VAT)
COMPARE
316L Stainless Steel 1 1
£47.67GBP / Each
£57.20GBP (incl VAT)
COMPARE
316L Stainless Steel 1-1/2 1-1/2
£48.76GBP / Each
£58.51GBP (incl VAT)
COMPARE
316L Stainless Steel 2 2
£55.26GBP / Each
£66.31GBP (incl VAT)