Masterflex® Union Fittings, Hose Barb, Y-Connector, Polyproylene
No tools needed for assembly
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Designed for quick and easy assembly with flexible tubing
Combine these simple, economical fittings with flexible tubing for use in your low pressure applications.
Ideal for applications such as pharmaceuticals, laboratory, industrial/manufacturing, water sampling, food processing.
£16.01 - £283.87GBP / Pkg of 10
£19.21 - £340.64GBP (incl VAT)
42 variations of this product are available.
See All Product Options Below
42 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in) Fitting Material
Availability Pricing
COMPARE
5/32 5/32 Polypropylene
£34.67GBP / Pkg of 20
£41.60GBP (incl VAT)
COMPARE
25/64 25/64 Polypropylene
£50.92GBP / Pkg of 20
£61.10GBP (incl VAT)
COMPARE
35/64 35/64 Polypropylene
£32.98GBP / Pkg of 10
£39.58GBP (incl VAT)
COMPARE
5/8 5/8 Polypropylene
£37.92GBP / Pkg of 10
£45.50GBP (incl VAT)
COMPARE
25/64 25/64 Polypropylene
£62.84GBP / Pkg of 20
£75.41GBP (incl VAT)
COMPARE
15/64 15/64 Polypropylene
£56.34GBP / Pkg of 20
£67.61GBP (incl VAT)
COMPARE
5/16 5/16 Polypropylene
£63.92GBP / Pkg of 20
£76.70GBP (incl VAT)
COMPARE
35/64 35/64 Polypropylene
£81.26GBP / Pkg of 20
£97.51GBP (incl VAT)
COMPARE
3/16 3/16 Polypropylene
£21.75GBP / Pkg of 5
£26.10GBP (incl VAT)
COMPARE
1/4 1/4 Polypropylene
£23.77GBP / Pkg of 5
£28.52GBP (incl VAT)
COMPARE
3/16 3/16 Polypropylene
£29.42GBP / Pkg of 10
£35.30GBP (incl VAT)
COMPARE
3/8 3/8 Polypropylene
£35.15GBP / Pkg of 10
£42.18GBP (incl VAT)
COMPARE
5/8 5/8 Polypropylene
£52.98GBP / Pkg of 10
£63.58GBP (incl VAT)
COMPARE
3/4 3/4 Polypropylene
£52.98GBP / Pkg of 10
£63.58GBP (incl VAT)
COMPARE
3/32 3/32 Polypropylene
£22.36GBP / Pkg of 10
£26.83GBP (incl VAT)
COMPARE
1/4 1/4 Polypropylene
£38.98GBP / Pkg of 10
£46.78GBP (incl VAT)
COMPARE
3/8 3/8 Polypropylene
£40.49GBP / Pkg of 10
£48.59GBP (incl VAT)
COMPARE
5/8 5/8 Polypropylene
£51.87GBP / Pkg of 10
£62.24GBP (incl VAT)
COMPARE
1/16 1/16 Polypropylene
£147.35GBP / Pkg of 100
£176.82GBP (incl VAT)
COMPARE
3/32 3/32 Polypropylene
£144.10GBP / Pkg of 100
£172.92GBP (incl VAT)
Showing 1 - 20 of 42