Masterflex® Union Fittings, Hose Barb, Tee, PVDF
No tools needed for assembly
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed tee fittings are designed for quick and easy assembly. Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
£24.88 - £372.71GBP / Pkg of 10
£29.86 - £447.25GBP (incl VAT)
20 variations of this product are available.
See All Product Options Below
20 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in) Fitting Material
Availability Pricing
COMPARE
3/16 3/16 PVDF
£35.11GBP / Pkg of 10
£42.13GBP (incl VAT)
COMPARE
3/8 3/8 PVDF
£35.75GBP / Pkg of 10
£42.90GBP (incl VAT)
COMPARE
1/2 1/2 PVDF
£39.00GBP / Pkg of 10
£46.80GBP (incl VAT)
COMPARE
5/8 5/8 PVDF
£45.51GBP / Pkg of 10
£54.61GBP (incl VAT)
COMPARE
3/4 3/4 PVDF
£54.17GBP / Pkg of 10
£65.00GBP (incl VAT)
COMPARE
1 1 PVDF
£68.26GBP / Pkg of 10
£81.91GBP (incl VAT)
COMPARE
5/16 5/16 PVDF
£36.84GBP / Pkg of 10
£44.21GBP (incl VAT)
COMPARE
3/16 3/16 PVDF
£36.84GBP / Pkg of 10
£44.21GBP (incl VAT)
COMPARE
5/16 5/16 PVDF
£132.18GBP / Pkg of 50
£158.62GBP (incl VAT)
COMPARE
3/8 3/8 Nylon
£206.94GBP / Pkg of 100
£248.33GBP (incl VAT)
COMPARE
1/4 1/4 PVDF
£33.59GBP / Pkg of 10
£40.31GBP (incl VAT)
COMPARE
3/8 3/8 PVDF
£36.84GBP / Pkg of 10
£44.21GBP (incl VAT)
COMPARE
1/2 1/2 PVDF
£37.92GBP / Pkg of 10
£45.50GBP (incl VAT)
COMPARE
5/8 5/8 PVDF
£46.59GBP / Pkg of 10
£55.91GBP (incl VAT)
COMPARE
3/4 3/4 PVDF
£35.75GBP / Pkg of 5
£42.90GBP (incl VAT)
COMPARE
1 1 PVDF
£41.17GBP / Pkg of 5
£49.40GBP (incl VAT)
COMPARE
1/4
£372.71GBP / Pkg
£447.25GBP (incl VAT)
COMPARE
1/16 1/16 PVDF
£24.88GBP / Pkg of 10
£29.86GBP (incl VAT)
COMPARE
1/8 1/8 PVDF
£27.29GBP / Pkg of 10
£32.75GBP (incl VAT)
COMPARE
3/32 3/32 PVDF
£35.87GBP / Pkg of 10
£43.04GBP (incl VAT)
Showing 1- 20 of 20