Masterflex® Transfer Tubing, Tygon® ND-100-80 Microbore, Non-DEHP

High resistance to kinking & collapse
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
Formulation: Tygon ND-100-80
Applications: General laboratory and pharmaceutical use, especially with precision injection or dispensing applications. Also excellent with low-flow peristaltic pump applications.
Characteristics: Extremely flexible. Non-toxic; nonpyrogenic; biocompatible. Crystal clear.
Certifications: Resin meets USP Class VI requirements, non-DEHP.
Temperature range: -31 to 185°F (-35 to 85°C)
Sterilize: Autoclave, ethylene oxide, or gamma irradiation.
£273.03 - £361.88GBP / Roll of 1
£327.64 - £434.26GBP (incl VAT)

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Length (ft) Tubing ID (in) Tubing OD (in)
Availability Pricing
COMPARE
500 1/100 1/32
£273.03GBP / Roll of 1
£327.64GBP (incl VAT)
COMPARE
500 1/50 1/16
£280.62GBP / Roll of 1
£336.74GBP (incl VAT)
COMPARE
500 1/32 3/32
£316.37GBP / Roll of 1
£379.64GBP (incl VAT)
COMPARE
500 1/25 5/64
£351.04GBP / Roll of 1
£421.25GBP (incl VAT)
COMPARE
500 1/20 3/32
£361.88GBP / Roll of 1
£434.26GBP (incl VAT)
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat