Masterflex® Transfer Tubing, PVC
Stop oxidation and bacteria problems
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
This tubing is extremely flexible and transparent and is resistant to oxidation, bacteria and a broad range of chemicals. PVC tubing meets FDA CFR Title 21 177.101.
£13.34 - £336.96GBP / Pkg of 1
£16.01 - £404.35GBP (incl VAT)
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
Compareitem specifications
Max Pressure (PSI) Tubing ID (in) Tubing OD (in)
Availability Pricing
COMPARE
1/8 1/4
£13.34GBP / Pkg of 1
£16.01GBP (incl VAT)
COMPARE
3/16 5/16
£18.19GBP / Pkg of 1
£21.83GBP (incl VAT)
COMPARE
1/4 3/8
£20.61GBP / Pkg of 1
£24.73GBP (incl VAT)
COMPARE
1/4 1/2
£46.59GBP / Pkg of 1
£55.91GBP (incl VAT)
COMPARE
5/16 7/16
£24.26GBP / Pkg of 1
£29.11GBP (incl VAT)
COMPARE
3/8 1/2
£27.89GBP / Pkg of 1
£33.47GBP (incl VAT)
COMPARE
3/8 5/8
Call For Price
COMPARE
1/2 5/8
£40.09GBP / Pkg of 1
£48.11GBP (incl VAT)
COMPARE
1/2 3/4
£55.55GBP / Pkg of 1
£66.66GBP (incl VAT)
COMPARE
5/8 7/8
Call For Price
COMPARE
3/4 1
£115.93GBP / Pkg of 1
£139.12GBP (incl VAT)
COMPARE
1 1 1/4
£150.60GBP / Pkg of 1
£180.72GBP (incl VAT)
COMPARE
1 1/4 1 3/4
£336.96GBP / Pkg of 1
£404.35GBP (incl VAT)
Showing 1- 13 of 13