Masterflex® Transfer Tubing, Metric Polytetrafluoroethylene (PTFE)
Withstands extreme temperatures
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
Applications: Demanding lab, chemical, electrical, mechanical, pharmaceutical, and automotive applications. Good for high-purity work. PTFE tubing withstands extreme temperatures.
Characteristics: Chemically inert, nontoxic, and virtually nonporous. Excellent crack- and stress-resistance. Low coefficient of friction. Translucent.
Resin certification: Resin is FDA-compliant (21 CFR 177.1550); meets USP 23 Class VI requirements. Tubing is manufactured according to GMP.
Temperature range: -400 to 500°F (-240 to 260°C)
Sterilize: With autoclave, ethylene oxide, or use dry heat.
£44.42 - £212.36GBP / Pkg of 1
£53.30 - £254.83GBP (incl VAT)
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (mm) Tubing OD (in) Tubing OD (mm)
Availability Pricing
COMPARE
6 9/32 7
£74.76GBP / Pkg of 1
£89.71GBP (incl VAT)
COMPARE
8 23/64 9
£79.09GBP / Pkg of 1
£94.91GBP (incl VAT)
COMPARE
10 15/32 12
£170.10GBP / Pkg of 1
£204.12GBP (incl VAT)
COMPARE
1.48 1/8 3
£44.42GBP / Pkg of 1
£53.30GBP (incl VAT)
COMPARE
2.48 5/32 4
£63.95GBP / Pkg of 1
£76.74GBP (incl VAT)
COMPARE
4.48 15/64 6
£84.45GBP / Pkg of 1
£101.34GBP (incl VAT)
COMPARE
6.22 5/16 8
£108.35GBP / Pkg of 1
£130.02GBP (incl VAT)
COMPARE
7.98 25/64 10
£183.11GBP / Pkg of 1
£219.73GBP (incl VAT)
COMPARE
9.46 15/32 12
£212.36GBP / Pkg of 1
£254.83GBP (incl VAT)
Showing 1- 9 of 9