Masterflex® Six-Channel Syringe Pumps
Six-channel pump can both withdraw and infuse
  • Flow rates up to 22.3 mL/min per syringe
  • Network up to 100 pumps from a single communications port on a computer
  • Economical pump can both withdraw and infuse
  • Pumps are capable of holding 41 commands, do unlimited repetitions and unlimited runtime
  • Built-i
Programming commands allow dispensing of a specified amount, automatic rates changes, direction changes, programmed pauses, external or user input, and sending or receive signals from other equipment. Non-volatile memory restores all setup parameters and
£2,455.14 - £2,698.92GBP / Each
£2,946.17 - £3,238.70GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy Power (VAC) Description
Availability Pricing
COMPARE
±1% 120 Six Channel Syringe Pump, 115 VAC
£2,698.92GBP / Each
£3,238.70GBP (incl VAT)
COMPARE
±1% 230 Six Channel Syringe Pump, 230v
£2,455.14GBP / Each
£2,946.17GBP (incl VAT)