Masterflex® Sanitary Clamp Fittings (Nylon)
Compatible with metal and plastic Tri-Clamp® fittings
  • Excellent chemical resistance; low leachable characteristics
  • Autoclavable
Constructed of nylon, these Tri-Clamp® sanitary clamps are perfect for single-use transfer lines in biopharma markets, plus a wide range of fluid transfer systems. These clamps ideal for securing your fluid transfer system without damaging the process lines.
£19.97 - £57.42GBP / Each
£23.96 - £68.90GBP (incl VAT)
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Fitting Material Connection Type Sanitary Clamp Size (Port 1) (in)
Availability Pricing
COMPARE
Nylon Sanitary 3/4
£19.97GBP / Each
£23.96GBP (incl VAT)
COMPARE
Nylon Sanitary 1-1/2
£26.21GBP / Each
£31.45GBP (incl VAT)
COMPARE
Nylon Sanitary 2
£53.48GBP / Each
£64.18GBP (incl VAT)
COMPARE
Nylon Sanitary 3
£52.18GBP / Each
£62.62GBP (incl VAT)
COMPARE
Nylon Sanitary 4
£57.42GBP / Each
£68.90GBP (incl VAT)