Masterflex® Replacement Pump Heads for Single-Use Pump Head System
Make the most of your single-use pump head systems
  • Deliver flow rates up to 500 mL/min at speeds up to 100 rpm
  • Steady, repeatable flow is ideal for dosing applications
  • Pump fluids with viscosities up to 70 cps; pressure up to 2.5 bar
  • Cost-effective solution for single-use / single-campaign fluid transfer
Note: Requires the mounting adapter included in the 07539-20 Single-Use Pump Head System
£98.65 - £539.42GBP / Pkg
£118.38 - £647.30GBP (incl VAT)
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Pulseless Disposable Pump Head for Masterflex L/S Drives; Single Pump Head
£98.65GBP / Each
£118.38GBP (incl VAT)
COMPARE
Pulseless Disposable Pump Head for Masterflex L/S Drives; Pack of Three Pump Heads
£277.85GBP / Pkg
£333.42GBP (incl VAT)
COMPARE
Pulseless Disposable Pump Head for Masterflex L/S Drives; Pack of Six Pump Heads
£539.42GBP / Pkg
£647.30GBP (incl VAT)