Masterflex® Reducer Fittings, Hose Barb, Straight (HDPE)
Best for low-pressure applications
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Temperture range -65 to 150°F (-53 to 65°C)
These economical straight barbed reducing fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.
£9.96 - £96.43GBP / Pkg of 5
£11.95 - £115.72GBP (incl VAT)
43 variations of this product are available.
See All Product Options Below
43 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in) Fitting Material
Availability Pricing
COMPARE
3/16 1/8 HDPE
£15.50GBP / Pkg of 10
£18.60GBP (incl VAT)
COMPARE
5/8 1/4 HDPE
£13.41GBP / Pkg of 10
£16.09GBP (incl VAT)
COMPARE
1 3/4 HDPE
£12.39GBP / Pkg of 5
£14.87GBP (incl VAT)
COMPARE
1 1/2 HDPE
£13.00GBP / Pkg of 5
£15.60GBP (incl VAT)
COMPARE
3/8 5/16 HDPE
£17.48GBP / Pkg of 10
£20.98GBP (incl VAT)
COMPARE
1/2 1/4 HDPE
£17.48GBP / Pkg of 10
£20.98GBP (incl VAT)
COMPARE
1/2 3/8 HDPE
£17.48GBP / Pkg of 10
£20.98GBP (incl VAT)
COMPARE
5/8 1/4 HDPE
£59.59GBP / Pkg of 100
£71.51GBP (incl VAT)
COMPARE
3/4 5/8 HDPE
£55.26GBP / Pkg of 50
£66.31GBP (incl VAT)
COMPARE
1/4 5/32 HDPE
£60.67GBP / Pkg of 100
£72.80GBP (incl VAT)
COMPARE
1/4 1/8 HDPE
£96.43GBP / Pkg of 100
£115.72GBP (incl VAT)
COMPARE
1/4 3/16 HDPE
£60.67GBP / Pkg of 100
£72.80GBP (incl VAT)
COMPARE
5/16 1/4 HDPE
£59.59GBP / Pkg of 100
£71.51GBP (incl VAT)
COMPARE
3/8 5/16 HDPE
£94.26GBP / Pkg of 100
£113.11GBP (incl VAT)
COMPARE
1/2 5/16 HDPE
£59.59GBP / Pkg of 100
£71.51GBP (incl VAT)
COMPARE
1/2 3/8 HDPE
£94.26GBP / Pkg of 100
£113.11GBP (incl VAT)
COMPARE
3/16 1/8 HDPE
£12.39GBP / Pkg of 10
£14.87GBP (incl VAT)
COMPARE
1/8 1/16 HDPE
£17.82GBP / Pkg of 10
£21.38GBP (incl VAT)
COMPARE
3/32 1/16 HDPE
£15.91GBP / Pkg of 10
£19.09GBP (incl VAT)
COMPARE
1/8 1/16 HDPE
£15.91GBP / Pkg of 10
£19.09GBP (incl VAT)
Showing 1 - 20 of 43