Masterflex® Reducer Fitting, Eccentric Sanitary Clamp, Straight
Meets FDA requirements and 3A sanitary standards
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Easily connects pipes or fittings of different types
  • Compatible with standard 304L fitting clamps
  • Ideal for pharmaceutical industry
These concentric reducers decrease in diameter towards the center thus forming a conical cylinder. The 316L stainless steel couplings are suitable for the pharmaceutical industry. They offer great chemical compatibility for use with corrosive acids and bases. Clamps and gaskets required for a complete system are sold separately.
£124.60 - £386.80GBP / Each
£149.52 - £464.16GBP (incl VAT)
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Fitting Material Sanitary Clamp Size (Port 1) (in) Sanitary Clamp Size (Port 2) (in)
Availability Pricing
COMPARE
316L Stainless Steel 1 3/4
£173.35GBP / Each
£208.02GBP (incl VAT)
COMPARE
316L Stainless Steel 1-1/2 1
£124.60GBP / Each
£149.52GBP (incl VAT)
COMPARE
316L Stainless Steel 2 1
£386.80GBP / Each
£464.16GBP (incl VAT)