Masterflex® Quick-Disconnect Fittings
Disconnect and reconnect your flow path with ease
  • Audible click lets you know the coupling is secure
  • Many hose barb sizes to choose from
  • Valved design allows you to cut off fluid flow when the fittings are disconnected
Use these fittings to quickly connect and disconnect from your flow line. Simply order a body (female half) and an insert (male half). Couplings are compatible for use with gases and liquids. Choose between acetal for a wide operating temperature range and greater durability or polypropylene (PP) for stronger chemical resistance and the ability to be gamma sterilized.
£3.91 - £8.55GBP / Each
£4.69 - £10.26GBP (incl VAT)
21 variations of this product are available.
See All Product Options Below
21 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Fitting Material Connection Type
Availability Pricing
COMPARE
1/16 Acetal Hose barb socket
£6.29GBP / Each
£7.55GBP (incl VAT)
COMPARE
1/8 Acetal Hose barb socket
£4.71GBP / Each
£5.65GBP (incl VAT)
COMPARE
3/16 Acetal Hose barb socket
£4.99GBP / Each
£5.99GBP (incl VAT)
COMPARE
1/4 Acetal Body
£4.71GBP / Each
£5.65GBP (incl VAT)
COMPARE
1/8 Polypropylene Hose barb socket
£8.17GBP / Each
£9.80GBP (incl VAT)
COMPARE
1/4 Polypropylene Hose barb socket
£8.17GBP / Each
£9.80GBP (incl VAT)
COMPARE
1/4 Acetal (POM) Quick-Disconnect x Barb
£4.67GBP / Each
£5.60GBP (incl VAT)
COMPARE
5/16 Acetal (POM) Quick-Disconnect x Barb
£4.67GBP / Each
£5.60GBP (incl VAT)
COMPARE
3/8 Acetal (POM) Quick-Disconnect x Barb
£4.67GBP / Each
£5.60GBP (incl VAT)
COMPARE
1/4 Polypropylene Quick-Disconnect x Barb
£8.55GBP / Each
£10.26GBP (incl VAT)
COMPARE
3/8 Polypropylene Quick-Disconnect x Barb
£8.55GBP / Each
£10.26GBP (incl VAT)
COMPARE
3/16 Acetal Hose barb plug
£6.10GBP / Each
£7.32GBP (incl VAT)
COMPARE
1/4 Acetal Hose barb plug
£4.48GBP / Each
£5.38GBP (incl VAT)
COMPARE
1/4 Acetal (POM) Quick-Disconnect x Barb
£3.91GBP / Each
£4.69GBP (incl VAT)
COMPARE
5/16 Acetal (POM) Quick-Disconnect x Barb
£3.91GBP / Each
£4.69GBP (incl VAT)
COMPARE
3/8 Acetal (POM) Quick-Disconnect x Barb
£3.91GBP / Each
£4.69GBP (incl VAT)
COMPARE
1/8 Polypropylene Hose barb plug
£6.14GBP / Each
£7.37GBP (incl VAT)
COMPARE
1/4 Polypropylene Hose barb plug
£5.88GBP / Each
£7.06GBP (incl VAT)
COMPARE
1/4 Polypropylene Quick-Disconnect x Barb
£5.97GBP / Each
£7.16GBP (incl VAT)
COMPARE
3/8 Polypropylene Quick-Disconnect x Barb
£6.58GBP / Each
£7.90GBP (incl VAT)
Showing 1 - 20 of 21