Masterflex® Press-In Plug Fittings, Hose Barb, Straight
Protect against contaminants in tubing lines
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Use these barbed press-in-plugs to keep open tubing lines or tubing assemblies free of foreign contaminants and ensure process purity. Plugs are manufactured from 100% virgin materials in a ISO Class 8 (formerly Class 100,000) environment and include full material traceability. Available in PVDF or nylon. All plugs are sterilizable by gamma irradiation; nylon plugs may also be autoclaved.
£2.34 - £20.05GBP / Pkg
£2.81 - £24.06GBP (incl VAT)
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Fitting Material
Availability Pricing
COMPARE
1/16 Nylon
£2.34GBP / Pkg of 10
£2.81GBP (incl VAT)
COMPARE
3/32 Nylon
£3.50GBP / Pkg of 10
£4.20GBP (incl VAT)
COMPARE
1/8 Nylon
£3.47GBP / Pkg of 10
£4.16GBP (incl VAT)
COMPARE
3/32 Nylon
£14.30GBP / Pkg
£17.16GBP (incl VAT)
COMPARE
1/16 PVDF
£15.31GBP / Pkg of 10
£18.37GBP (incl VAT)
COMPARE
1/8 PVDF
£14.60GBP / Pkg of 10
£17.52GBP (incl VAT)
COMPARE
5/32 PVDF
£17.73GBP / Pkg of 10
£21.28GBP (incl VAT)
COMPARE
1/16
£15.11GBP / Pkg of 10
£18.13GBP (incl VAT)
COMPARE
1/16 HDPE
£14.30GBP / Pkg
£17.16GBP (incl VAT)
COMPARE
1/16 PVDF
£16.92GBP / Pkg
£20.30GBP (incl VAT)
COMPARE
1/8 Nylon
£12.99GBP / Pkg of 10
£15.59GBP (incl VAT)
COMPARE
1/8 HDPE
£12.99GBP / Pkg
£15.59GBP (incl VAT)
COMPARE
1/8 PVDF
£17.42GBP / Pkg of 10
£20.90GBP (incl VAT)
COMPARE
3/16 PVDF
£20.05GBP / Pkg of 10
£24.06GBP (incl VAT)
COMPARE
1/16 Polypropylene
£15.11GBP / Pkg
£18.13GBP (incl VAT)
COMPARE
1/8 Polypropylene
£14.30GBP / Pkg
£17.16GBP (incl VAT)
Showing 1- 16 of 16