Masterflex® L/S® Standard Pump Heads for Precision Tubing
Ideal for fluid transfer applications requiring high accuracy and repeatability
  • Standard heads are precision molded to accommodate a single size of tubing
  • Pump head is compact and simple in design with same side tubing entry/exit—perfect for space-limited applications
  • Standard heads are compact and easy to integrate into your OEM applications
  • Stack up to four heads depending on drive recommendations
This Standard pump head accepts precision tubing— ensures optimal performance from your Masterflex pump. Select a pump head for one of the following precision tubing sizes: L/S 13, L/S 14, L/S 16, L/S 17, L/S 18.
£189.16 - £305.91GBP / Each
£226.99 - £367.09GBP (incl VAT)
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Housing Material Rotor Material Tubing sizes accepted
Availability Pricing
COMPARE
Polycarbonate (PC) Cold-rolled steel (CRS) L/S® 13
£189.16GBP / Each
£226.99GBP (incl VAT)
COMPARE
Polycarbonate (PC) Stainless Steel L/S® 13
£275.14GBP / Each
£330.17GBP (incl VAT)
COMPARE
Polyphenylene Sulfide (PPS) Stainless Steel L/S® 13
£305.91GBP / Each
£367.09GBP (incl VAT)
COMPARE
Polycarbonate (PC) Cold-rolled Steel L/S® 14
£189.16GBP / Each
£226.99GBP (incl VAT)
COMPARE
Polycarbonate (PC) Stainless Steel L/S® 14
£275.14GBP / Each
£330.17GBP (incl VAT)
COMPARE
Polyphenylene Sulfide (PPS) Stainless Steel L/S® 14
£305.91GBP / Each
£367.09GBP (incl VAT)
COMPARE
Polycarbonate (PC) Cold-rolled Steel L/S® 16
£189.16GBP / Each
£226.99GBP (incl VAT)
COMPARE
Polycarbonate (PC) Stainless Steel L/S® 16
£275.14GBP / Each
£330.17GBP (incl VAT)
COMPARE
Polyphenylene Sulfide (PPS) Stainless Steel L/S® 16
£305.91GBP / Each
£367.09GBP (incl VAT)
COMPARE
Polycarbonate (PC) Cold-rolled Steel L/S® 17
£189.16GBP / Each
£226.99GBP (incl VAT)
COMPARE
Polycarbonate (PC) Stainless Steel L/S® 17
£275.14GBP / Each
£330.17GBP (incl VAT)
COMPARE
Polyphenylene Sulfide (PPS) Stainless Steel L/S® 17
£305.91GBP / Each
£367.09GBP (incl VAT)
COMPARE
Polycarbonate (PC) Cold-rolled Steel L/S® 18
£189.16GBP / Each
£226.99GBP (incl VAT)
COMPARE
Polycarbonate (PC) Stainless Steel L/S® 18
£275.14GBP / Each
£330.17GBP (incl VAT)
COMPARE
Polyphenylene Sulfide (PPS) Stainless Steel L/S® 18
£305.91GBP / Each
£367.09GBP (incl VAT)
Showing 1- 15 of 15