Masterflex® L/S® Standard Pump Heads for High-Performance Precision Tubing
Ideal for fluid transfer applications requiring high accuracy and repeatability
  • Standard heads are precision molded to accommodate a single size of tubing
  • Pump head is compact and simple in design with same side tubing entry/exit—perfect for space-limited applications
  • Standard heads are compact and easy to integrate into your OEM applications
  • Stack up to four heads depending on drive recommendations
This Standard pump head accepts high-performance precision tubing— choose these pump heads for better pressure generation, suction lift, viscous fluid transfer, and longer tubing life. Select a pump head for one of the following high-performance tubing sizes: L/S 15, L/S 24, L/S 35, L/S 36.
£189.16 - £314.96GBP / Each
£226.99 - £377.95GBP (incl VAT)
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Housing Material Rotor Material Tubing sizes accepted
Availability Pricing
COMPARE
Polycarbonate (PC) Cold-rolled Steel L/S® 15
£189.16GBP / Each
£226.99GBP (incl VAT)
COMPARE
Polycarbonate (PC) Stainless Steel L/S® 15
£275.14GBP / Each
£330.17GBP (incl VAT)
COMPARE
Polyphenylene Sulfide (PPS) Stainless Steel L/S® 15
£305.91GBP / Each
£367.09GBP (incl VAT)
COMPARE
Polycarbonate (PC) Cold-rolled Steel L/S® 24
£189.16GBP / Each
£226.99GBP (incl VAT)
COMPARE
Polycarbonate (PC) Stainless Steel L/S® 24
£275.14GBP / Each
£330.17GBP (incl VAT)
COMPARE
Polyphenylene Sulfide (PPS) Stainless Steel L/S® 24
£305.91GBP / Each
£367.09GBP (incl VAT)
COMPARE
Polycarbonate (PC) Cold-rolled Steel L/S® 35
£189.16GBP / Each
£226.99GBP (incl VAT)
COMPARE
Polycarbonate (PC) Stainless Steel L/S® 35
£275.14GBP / Each
£330.17GBP (incl VAT)
COMPARE
Polycarbonate (PC) Cold-rolled Steel L/S® 36
£255.23GBP / Each
£306.28GBP (incl VAT)
COMPARE
Polycarbonate (PC) Stainless Steel L/S® 36
£314.96GBP / Each
£377.95GBP (incl VAT)
Showing 1- 10 of 10