Masterflex® L/S® Standard Digital Pump Systems
Designed with maintenance-free high-accuracy motor and a graphical interface—makes setup and operation easier than ever!
  • Digital interface with graphical LCD shows pump performance and operating modes—continuous run, timed dispense, copy dispense, or volume dispense
  • Select your preferred language from the user-friendly menu for programming and operation
  • Dispense by Volume in mL (0.001 to 9,999 mL), L, or gal; Copy—1 to 9,999 dispense cycles; or Time—time interval of on/off cycles (1 sec to 99 hr, 59 min, 59 sec per cycle)Antidrip function ensures dispensing accuracy
  • Brushless, maintenance-free motor offers ±0.1% speed control accuracy with a 6000 to 1 turndown ratio
  • Models with open-head sensor stop drive motor when pump head is opened
These precision drives offer analog remote control options and simple programming—ideal for automated process applications. Drives feature a programmable dispense interval that lets you set the delay between dispense cycles for convenient, automated dispensing. Programmed calibration ensures dispense and display accuracy—drive stores one calibration value per tubing size.
Program the drive via the simple membrane keypad; keypad lock/unlock feature protects settings. Motor is reversible so you can purge tubing before or after pumping and pump fluid in either direction.
£2,778.53 - £3,846.50GBP / Each
£3,334.24 - £4,615.80GBP (incl VAT)
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate Per Channel (mL/min) Min Flow Rate Per Channel (mL/min) Pump Head
Availability Pricing
COMPARE
18 0.0034 07519-06 (L/S® Multichannel Cartridge Pump)
£3,846.50GBP / Each
£4,615.80GBP (incl VAT)
COMPARE
12 0.0024 07519-20 (L/S® Multichannel Cartridge Pump)
£3,276.31GBP / Each
£3,931.57GBP (incl VAT)
COMPARE
480 0.08 77800-60 (L/S® Easy-Load 3 Pump Head)
£2,778.53GBP / Each
£3,334.24GBP (incl VAT)
COMPARE
1700 0.28 77200-62 (L/S® Easy-Load II Pump Head)
£2,837.36GBP / Each
£3,404.83GBP (incl VAT)
COMPARE
480 0.08 77200-30 (L/S® Easy-Load® II Head with Open-Head Sensor)
£3,090.78GBP / Each
£3,708.94GBP (incl VAT)
COMPARE
1700 0.28 77200-32 (L/S® Easy-Load® II Head with Open-Head Sensor)
£3,095.30GBP / Each
£3,714.36GBP (incl VAT)
COMPARE
280 0.047 07535-04 (L/S® 4-Channel Pump Head)
£3,371.35GBP / Each
£4,045.62GBP (incl VAT)
Showing 1- 7 of 7