Masterflex® L/S® Precision Tygon Chemical (2001) Pump Tubing
Compatible with aggressive chemicals
  • Outstanding chemical resistance
  • Cost-effective alternative to fluoropolymers or fluoroelastomers
  • FDA compliant
  • NOTE: Tygon® Chemical 2001 pump tubing is recommended for use in Easy-Load® and Easy-Load® II pump heads only.
Applications: Chemical, food, beverage, and pharmaceutical applications.
Characteristics: Excellent chemical resistance, low extractability, low gas permeability, plasticizer free. Translucent.
L/S® Precision Tygon® Chemical pump tubing offers excellent chemical compatibility in a clear, flexible peristaltic pump tube. This material is compatible with many challenging fluids including concentrated acids, strong bases, and organic solvents. Transparent for easy flow monitoring and plasticizer-free to protect fluid purity.
Masterflex® Tygon Chemical pump tubing is specially engineered and precision extruded for optimum performance in Masterflex® and other peristaltic pumps. Factory tested and optically inspected, it is available in all Masterflex® L/S® Precision and High-Performance Precision tube sizes, and all I/P® Precision tube sizes.
For interactive chemical compatibility charts, click on the Technical Info tab at the top of the page.
£63.60 - £165.36GBP / Pkg of 1
£76.32 - £198.43GBP (incl VAT)
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Hose Barb Size (in) L/S Masterflex tubing size Max Flow Rate (mL/min)
Availability Pricing
COMPARE
1/16 13 36
£63.60GBP / Pkg of 1
£76.32GBP (incl VAT)
COMPARE
1/16 14 130
£65.72GBP / Pkg of 1
£78.86GBP (incl VAT)
COMPARE
1/8 16 480
£100.70GBP / Pkg of 1
£120.84GBP (incl VAT)
COMPARE
1/4 17 1700
£125.08GBP / Pkg of 1
£150.10GBP (incl VAT)
COMPARE
3/8 18 2300
£165.36GBP / Pkg of 1
£198.43GBP (incl VAT)
COMPARE
3/16 25 1000
£121.90GBP / Pkg of 1
£146.28GBP (incl VAT)