Masterflex® L/S® Precision Pump Tubing, PharMed® BPT, Double-Y Elements
Protect your light-sensitive samples
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
Applications: Cell and tissue culture work, pharmaceutical, light-sensitive samples
Characteristics: Nontoxic, nonhemolytic, extremely low permeability, resists acids, bases, and oxidizing agents; heat-sealable, bondable, formable, opaque beige
£37.54 - £52.97GBP / Each
£45.05 - £63.56GBP (incl VAT)
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Hose Barb Size (in) L/S Masterflex tubing size Description
Availability Pricing
COMPARE
1/16 13 L/S® Precision Pump Tubing, PharMed® BPT Plus PVDF Fitting, Double Y, L/S 13
£37.54GBP / Each
£45.05GBP (incl VAT)
COMPARE
1/16 14 L/S® Precision Pump Tubing, PharMed® BPT Plus PVDF Fitting, Double Y, L/S 14
£37.54GBP / Each
£45.05GBP (incl VAT)
COMPARE
1/8 16 L/S® Precision Pump Tubing, PharMed® BPT Plus PVDF Fitting, Double Y, L/S 16
£37.54GBP / Each
£45.05GBP (incl VAT)
COMPARE
1/4 17 L/S® Precision Pump Tubing, PharMed® BPT Plus PVDF Fitting, Double Y, L/S 17
£41.52GBP / Each
£49.82GBP (incl VAT)
COMPARE
3/8 18 L/S® Precision Pump Tubing, PharMed® BPT Plus PVDF Fitting, Double Y, L/S 18
£52.97GBP / Each
£63.56GBP (incl VAT)
COMPARE
3/16 25 L/S® Precision Pump Tubing, PharMed® BPT Plus PVDF Fitting, Double Y, L/S 25
£41.52GBP / Each
£49.82GBP (incl VAT)