Masterflex® L/S® Multichannel Pump Heads
Get low-pulsation synchronous flow in multiple channels without cartridges
  • Flow range 0.045 to 2300 mL/min per channel— flow rate depends on drive rpm and tubing size
  • Stainless steel (SS) rotor / roller for greater durability and chemical resistance
  • Stackable two-, four-, or eight-channel heads for up to 24 channels
  • Simple tubing installation—lLoad the tubing, latch the occlusion bed, and run the pump
The L/S® multichannel pump heads are designed with machined stainless steel (SS) and anodized aluminum with SS rotor, bearings, and rollers for reliable, continuous-duty operation. Choose pump heads for microbore tubing, L/S Precision tubing, or L/S High-Performance Precision tubing based on your application requirements. Compatible with L/S® drives that accept two or more pump heads and have a 1/10 hp minimum.
£921.35 - £1,301.48GBP / Each
£1,105.62 - £1,561.78GBP (incl VAT)
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate Per Channel (mL/min) Number of Channels Tubing sizes accepted
Availability Pricing
COMPARE
200 4 Two-stop microbore
£921.35GBP / Each
£1,105.62GBP (incl VAT)
COMPARE
200 8 Two-stop microbore
£1,301.48GBP / Each
£1,561.78GBP (incl VAT)
COMPARE
280 4 Two-stop L/S® 13, L/S 14, L/S 16
£921.35GBP / Each
£1,105.62GBP (incl VAT)
COMPARE
280 8 Two-stop L/S® 13, L/S 14, L/S 16
£1,301.48GBP / Each
£1,561.78GBP (incl VAT)
COMPARE
2300 2 Two-stop L/S® 15, L/S 24, L/S 35
£921.35GBP / Each
£1,105.62GBP (incl VAT)
COMPARE
2300 4 Two-stop L/S® 15, L/S 24, L/S 35
£1,301.48GBP / Each
£1,561.78GBP (incl VAT)