Masterflex® L/S® High-Performance Pump Heads
All-new patented design offers faster tubing changes and improved ergonomics
  • Tubing occlusion is automatic and optimized when pump head is closed and latched – load tubing in 15 seconds or less
  • Flow rates up to 3400 mL/min—flow rate depends on drive rpm, pump head, and tubing size
  • Pressure up to 150 psi (10.3 bar) with L/S high-pressure (HP) pump tubing
  • Reinforced nylon housing and occlusion bed with stainless steel rotor/roller assembly
  • Compatible with all 1/10 hp L/S drives that accept two or more pump heads
  • Compact design for efficient use of space
The L/S® High-Performance pump heads offer better suction lift, viscous fluid transfer, and longer tubing life. Tubing enters and exits the same side of the pump head for easy connection to tubing inlet and outlet. Heads are not compatible with the 07554-series L/S Economy Console drives.
£534.88 - £685.12GBP / Each
£641.86 - £822.14GBP (incl VAT)
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate (mL/min) Open-Head Sensor Description
Availability Pricing
COMPARE
2812 Yes L/S® High-Performance Head with Open-Head Sensor for Precision Tubing
£598.23GBP / Each
£717.88GBP (incl VAT)
COMPARE
3400 Yes L/S® High-Performance Head with Open-Head Sensor for High-Performance Precision Tubing
£685.12GBP / Each
£822.14GBP (incl VAT)
COMPARE
2812 No L/S® High-Performance Head for Precision Tubing
£534.88GBP / Each
£641.86GBP (incl VAT)
COMPARE
3400 No L/S® High-Performance Head for High-Performance Precision Tubing
£536.86GBP / Each
£644.23GBP (incl VAT)
COMPARE
2812 No L/S® High-Performance Head for Precision Tubing, ATEX-approved
£620.52GBP / Each
£744.62GBP (incl VAT)
COMPARE
3400 No L/S® High-Performance Head for High-Performance Precision Tubing, ATEX-approved
£620.52GBP / Each
£744.62GBP (incl VAT)