Masterflex® L/S® Easy-Load® Pump Heads for Precision Tubing
Save time with fast tubing changes and reduced maintenance
  • New design and automatic tubing retention allow for multiple tubing sizes—change tubing without removing head from drive
  • Rotor is constructed of stainless steel (SS) for enhanced durability
  • Pump heads are stackable for multichannel applications
  • Compact design for efficient use of space
  • Suitable for use with L/S® Precision pump tubing sizes
This Easy-Load® pump head accepts precision tubing— ensures optimal performance from your Masterflex pump. Select a pump head for one of the following precision tubing sizes: L/S 13, L/S 14, L/S 16, L/S 25, L/S 17, L/S 18.
£336.68 - £386.36GBP / Each
£404.02 - £463.63GBP (incl VAT)
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate (mL/min) Open-Head Sensor Tubing sizes accepted
Availability Pricing
COMPARE
2300 No L/S® 13, L/S 14, L/S 16, L/S 25, L/S 17, L/S 18
£336.68GBP / Each
£404.02GBP (incl VAT)
COMPARE
2300 Yes L/S® 13, L/S 14, L/S 16, L/S 25, L/S 17, L/S 18
£386.36GBP / Each
£463.63GBP (incl VAT)
COMPARE
900 No L/S® 13, L/S 14, L/S 16, L/S 25
£360.21GBP / Each
£432.25GBP (incl VAT)