Masterflex® Correlated Variable-Area Flowmeters, High-Res Valve, High-Flow, 150-mm
Don’t sacrifice suitability for precision
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate│Use with a variety of media for laboratory and industrial applications
Aluminum is economical and good for general use with noncorrosive gases and liquids. Brass is economical and good for use with water. Stainless steel withstands higher temperature and pressure and features excellent chemical compatibility. Valved meters have valves mounted at the bottom/inlet, typically used for positive pressure with liquids and gases. The valve can be reconfigured to mount at the top/outlet, often for vacuum applications. All correlated flowmeters come with data sheets for water and air at standard temperature and pressure (STP).
£201.52 - £352.13GBP / Each
£241.82 - £422.56GBP (incl VAT)
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
Compareitem specifications
Material Fitting Process Connection (NPT(F)) Description
Availability Pricing
COMPARE
PTFE and Kel-F® 0 150-mm Correlated Flowmeter, with valve 806 mL/min water flow rate
£328.29GBP / Each
£393.95GBP (incl VAT)
COMPARE
Aluminum 0 150-mm Correlated Flowmeter, hi-res valve, aluminum, 23.742 LPM air
£206.94GBP / Each
£248.33GBP (incl VAT)
COMPARE
Aluminum 0 150-mm Correlated Flowmeter, hi-res valve, aluminum, 45.227 LPM air
£201.52GBP / Each
£241.82GBP (incl VAT)
COMPARE
Aluminum 0 150-mm Correlated Flowmeter, hi-res valve, aluminum, 66.346 LPM air
£206.94GBP / Each
£248.33GBP (incl VAT)
COMPARE
PTFE and Kel-F® 0 150-mm Correlated Flowmeter, hi-res valve, PTFE, 22.536 LPM air
£328.29GBP / Each
£393.95GBP (incl VAT)
COMPARE
316 SS 0 150-mm Correlated Flowmeter, hi-res valve, 316SS, 23.742 LPM air
£352.13GBP / Each
£422.56GBP (incl VAT)
COMPARE
316 SS 0 150-mm Correlated Flowmeter, hi-res valve, 316SS, 45.227 LPM air
£352.13GBP / Each
£422.56GBP (incl VAT)
COMPARE
316 SS 0 150-mm Correlated Flowmeter, hi-res valve, 316SS, 66.346 LPM air
£352.13GBP / Each
£422.56GBP (incl VAT)
COMPARE
Chrome-plated brass 0 150-mm Correlated Flowmeter, hi-res valve, brass, 23.742 LPM air
£221.03GBP / Each
£265.24GBP (incl VAT)
COMPARE
Chrome-plated brass 0 150-mm Correlated Flowmeter, hi-res valve, brass, 45.227 LPM air
£221.03GBP / Each
£265.24GBP (incl VAT)
COMPARE
Chrome-plated brass 0 150-mm Correlated Flowmeter, hi-res valve, brass, 66.346 LPM air
£221.03GBP / Each
£265.24GBP (incl VAT)
Showing 1- 11 of 11