Masterflex® Console Gear Pump for MicroPump A-Mount Pump Heads
Simple variable speed control handles many fluid handling applications
  • 100:1 speed-control range controls flow over a wide range
  • Maximum continuous-duty flow rates possible with Micropump® A-mount pump heads
  • Simple operation with single-turn speed control
  • Separate power switch enables you to maintain speed setting when turning drive on/off; front-panel LED indicates power on
  • Remote control of start/stop via DB9 connector on back of drive
Compact, cost-effective drive offers simple variable speed control for many fluid handling applications from general transfer to filtration to chemical injection. Couple with an A-mount gear pump head for pulseless fluid flow.
£1,003.26 - £1,080.22GBP / Each
£1,203.91 - £1,296.26GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (mL/min) Max Motor RPM (rpm) Power (VAC)
Availability Pricing
COMPARE
5850 5000 115
£1,003.26GBP / Each
£1,203.91GBP (incl VAT)
COMPARE
5850 5000 230
£1,080.22GBP / Each
£1,296.26GBP (incl VAT)