Masterflex® Close-Coupled Industrial Gear Pump Heads
Rugged corrosion-resistant bronze construction provides long service life
  • Easy assembly to NEMA and IEC motor frames
These gear pump heads are suitable for all liquids that are compatible with bronze—most common liquids are water, oil, and mild chemicals (pH from 4 to 11).
Choose a gear pump head to mount to your own motor (adapters for NEMA or IEC motors are sold separately below) or select a complete system with the pump head conveniently mounted to a standard (NEMA 56C) TEFC motor.
£295.05 - £414.75GBP / Each
£354.06 - £497.70GBP (incl VAT)
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate (GPM) Wetted Parts Description
Availability Pricing
COMPARE
1 Stainless Steel Bronze Close-Coupled Industrial Gear Pump Head; 1.0 GPM
£363.89GBP / Each
£436.67GBP (incl VAT)
COMPARE
2.1 Stainless Steel Bronze Close-Coupled Industrial Gear Pump Head; 2.1 GPM
£295.05GBP / Each
£354.06GBP (incl VAT)
COMPARE
4.4 Stainless Steel Bronze Close-Coupled Industrial Gear Pump Head; 4.4 GPM
£414.75GBP / Each
£497.70GBP (incl VAT)
COMPARE
1 Stainless Steel Bronze Close-Coupled Industrial Gear Pump Head; Relief Valve, 1.0 GPM
£409.50GBP / Each
£491.40GBP (incl VAT)
COMPARE
2.1 Stainless Steel Bronze Close-Coupled Industrial Gear Pump Head; Relief Valve, 2.1 GPM
£324.45GBP / Each
£389.34GBP (incl VAT)
COMPARE
4.4 Stainless Steel Bronze Close-Coupled Industrial Gear Pump Head; Relief Valve, 4.4 GPM
£369.18GBP / Each
£443.02GBP (incl VAT)