Manual Vacuum Valves
These manual vacuum valves are available as either a needle or isolation valve. The vacuum needle valve model is ideal for fine control or gas bleed. The vacuum isolation valve model is ideal for closing off sections of your vacuum system. An indicator displays open or closed position.
£391.79GBP / Each
£470.15GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Edwards C333-55-000 Vacuum isolation valve, NW 25 connections WZ-79302-60 Mfr # C333-55-000
COMPARE
Vacuum isolation valve, NW 25 connections
£391.79GBP / Each
£470.15GBP (incl VAT)