This product is now discontinued, View Related Products
Manning Peristaltic Sampling Pump
Double-wall, insulated base keeps samples and ice cooler longer
  • Meets EPA requirement for transport velocity of 2 ft/sec
  • Purge function clears air or obstructions from tubing that interfere with sample collecting, or clears intake line after sampling is complete
  • Changing tubing is safe and easy—safety kill switch automatically stops pump head rotation when lid is open
  • Sample collects in 4-gallon bottle; “bottle full” sensor automatically stops pump when full
  • Continuity liquid sensor ensures sample volume matches programmed volume
Advanced programming options include hydrologic level event mode (storm-water sampling), intake fault alert, intake line purge and rinse, sequential sampler start, and more. "Hot keys" let you quickly select the program you need—without scrolling through long menus.
Sampler base is made of impact and corrosion resistant ABS plastic with stainless steel hardware. The NEMA 4X, 6P controller and hermetically sealed keypad make the pump weatherproof. The two-line, alphanumeric backlit LCD is easy to read and program, even in low light.
What's included: sampling pump, 4-gallon polyethylene bottle with case, rechargeable battery, 115 VAC battery charger, 3/8" ID x 5/8" OD silicone tubing, 10-ft PVC tubing (3/8" ID), and PVC strainer.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Length (in) Material Sample Volume
Availability Pricing
Manning MS889040 Suspension Harness for Peristalitic and Vacuum Samplers WZ-99240-52 Mfr # MS889040
COMPARE
Call For Price
Manning PST8B2A2A4B2 Peristaltic Sampling Pump, 4 gal max volume WZ-99242-00 Mfr # PST8B2A2A4B2