M-System Limit Alarms
Monitor a range of alarm conditions
  • Thumbwheel switch adjustments
  • Only 1.2 inch (29.5mm) wide
  • Front LCD display indicating values in scaled engineering unit range is used to program the module
  • Quad or dual trip
  • Program Lockout function
  • Front-panel configurable
£230.91GBP / Each
£277.09GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Load Rating Max Current (amps) Relay Type
Availability Pricing
M-System M2SED-A13-R/UL Dc Input Limit Alarm Thin Profile 4-20 Ma Dual Spdt WZ-68467-64 Mfr # M2SED-A13-R/UL
COMPARE
SPDT
£230.91GBP / Each
£277.09GBP (incl VAT)