Lowboy Low-Profile Carboys
Space-saving design is ideal for shelves
  • Removable spigot provides quick dispensing
  • Easily attach tubing to spigot
Space-saving, low-profile carboys fit nicely into tight spaces. Removable fast-action PP spigot accepts 5/8" ID tubing. Use the included HDPE tubing adapter to connect tubing with ¼" to 5/16" ID.
£274.65GBP / Each
£329.58GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (L) Cap / Stopper Size Material
Availability Pricing
Thermo Scientific Nalgene 2323-0008 Lowboy Low-Profile HDPE Carboy, 8 L WZ-06059-05 Mfr # 2323-0008
COMPARE
8 63 mm High Density Polyethyene (HDPE)
£274.65GBP / Each
£329.58GBP (incl VAT)