This product is now discontinued, View Related Products
Loop Leader Series Loop-Powered Rate/Totalizers
Intrinsically safe model for use in just about any hazardous area.
  • 5-digit backlit display with bargraph
  • Open collector alarm or pulse output
  • HART protocol transparent
The loop-powered rate/totalizers can display rate up to five digits and total up to eight digits using the total overflow feature. The display can be programmed to automatically or manually toggle between rate and total. Standard features include a full 5-digit backlit display with bargraph, on-screen custom engineering units, and max/min display capabilities.
Square root and ptrogrammable exponent functions are standard features on these meters. These indicators can display flow rate and total from a DP transmitter or open channel flow in weirs and flumes. Programming is easy through the fron panel buttons, with no pots or jumpers to deal with, and can be scaled without a signal source.
The Loop Leader Series meters go just about anywhere. The PD689, with FM, CSA and ATEX certifications, can be installed in just about any hazardous area.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
DIN Size Input Type
Availability Pricing
Precision Digital PD684-0K1 Process meter, general purpose WZ-61600-02 Mfr # PD684-0K1
COMPARE
1/8\"
Call For Price
Precision Digital PD689-0K1 Process meter, hazardous area WZ-61600-03 Mfr # PD689-0K1
COMPARE
1/8\"